siRNA Details
| Gene Symbol: Olr1314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000468.1 | NP_001000468.1 | 904 | |||
| Gene Symbol: LOC103693060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766126.1 | XP_008764348.1 | 1 | 904 | ||

Olr1314
Genomic Map