siRNA Details
| Gene Symbol: Olr1315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000469.1 | NP_001000469.1 | 792 | |||
| Gene Symbol: LOC103690273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766127.1 | XP_008764349.1 | 1 | 792 | ||

Olr1315
Genomic Map