siRNA Details
| Gene Symbol: Olr1323 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000472.1 | NP_001000472.1 | 1 | 518 | ||
| Gene Symbol: LOC100362319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243144.2 | XP_006243206.2 | 1 | 518 | ||

Olr1323
Genomic Map