siRNA Details
| Gene Symbol: KIF14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305792.1 | NP_001292721.1 | 1 | 568 | ||
| NM_014875.2 | NP_055690.1 | 2 | 892 | |||
| XM_011510231.2 | XP_011508533.1 | 3 | 986 | |||
| XM_011510232.2 | XP_011508534.1 | 2 | 648 | |||
| XM_011510233.2 | XP_011508535.1 | 3 | 779 | |||
| XM_011510236.1 | XP_011508538.1 | 2 | 863 | |||
| XM_017003005.1 | XP_016858494.1 | 1 | 1069 | |||
| XM_017003006.1 | XP_016858495.1 | 2 | 677 | |||
| GenBank mRNA | BC113742.1 | - | 1 | 531 | 8322794 | |
| D26361.1 | - | 2 | 892 | |||

KIF14
Genomic Map