siRNA Details
| Gene Symbol: Olr1638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000834.1 | NP_001000834.1 | 1 | 274 | ||
| XM_008770559.1 | XP_008768781.1 | 2 | 857 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1638| Gene Symbol: Olr1638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000834.1 | NP_001000834.1 | 1 | 274 | ||
| XM_008770559.1 | XP_008768781.1 | 2 | 857 | |||