siRNA Details
| Gene Symbol: BC051142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001177.2 | NP_001001177.2 | 20 | 1539 | ||
| NM_001163855.1 | NP_001157327.1 | 24 | 1214 | |||
| GenBank mRNA | AK077077.1 | - | 20 | 1526 | ||
| AK133031.1 | - | 19 | 1372 | |||
| BC051142.1 | - | 24 | 1214 | 6740688 | ||

BC051142
Genomic Map