siRNA Details
| Gene Symbol: Zfp941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001180.2 | NP_001001180.2 | 3 | 2277 | ||
| GenBank mRNA | BC066028.1 | - | 3 | 2231 | 5702088 | |
| BC068250.1 | - | 3 | 2247 | 30018777 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp941| Gene Symbol: Zfp941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001180.2 | NP_001001180.2 | 3 | 2277 | ||
| GenBank mRNA | BC066028.1 | - | 3 | 2231 | 5702088 | |
| BC068250.1 | - | 3 | 2247 | 30018777 | ||