siRNA Details
| Gene Symbol: Tmem204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001183.1 | NP_001001183.1 | 3 | 1720 | ||
| GenBank mRNA | AK146008.1 | - | 3 | 1788 | ||
| BC019551.1 | - | 3 | 1734 | |||
| BC043726.1 | - | 3 | 1720 | |||
| BC054438.1 | - | 3 | 1720 | 6390142 | ||

Tmem204
Genomic Map