siRNA Details
Gene Symbol: Gm4759 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041093.1 | - | 3 | 1460 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm4759 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041093.1 | - | 3 | 1460 |