siRNA Details
| Gene Symbol: Hexdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001333.2 | NP_001001333.2 | 13 | 2044 | ||
| NM_001146073.1 | NP_001139545.1 | 12 | 1849 | |||
| GenBank mRNA | AK154237.1 | - | 13 | 2018 | ||
| BC056487.1 | - | 9 | 3814 | |||
| BC069960.1 | - | 12 | 1801 | 5007884 | ||

Hexdc
Genomic Map