siRNA Details
| Gene Symbol: Hexdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001333.2 | NP_001001333.2 | 12 | 1677 | ||
| NM_001146073.1 | NP_001139545.1 | 11 | 1482 | |||
| GenBank mRNA | AK154237.1 | - | 12 | 1651 | ||
| BC056487.1 | - | 8 | 3447 | |||
| BC069960.1 | - | 11 | 1434 | 5007884 | ||
| FM204885.1 | - | 11 | 1214 | |||
| FM204886.1 | - | 11 | 1433 | |||

Hexdc
Genomic Map