siRNA Details
| Gene Symbol: Plekha8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001335.2 | NP_001001335.1 | 4 | 591 | ||
| NM_001164361.1 | NP_001157833.1 | 4 | 686 | |||
| GenBank mRNA | AK078040.1 | - | 4 | 687 | ||
| AK170592.1 | - | 4 | 453 | |||
| BC052360.1 | - | 4 | 591 | 6704789 | ||

Plekha8
Genomic Map