siRNA Details
| Gene Symbol: Trpv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001445.2 | NP_001001445.1 | 11 | 1943 | ||
| GenBank mRNA | AB180097.1 | - | 10 | 1691 | ||
| AJ620495.1 | - | 10 | 1731 | |||
| AK051315.1 | - | 11 | 1801 | |||
| AY445519.1 | - | 10 | 1691 | |||
| AY452083.1 | - | 10 | 1724 | |||
| AY452084.1 | - | 10 | 1694 | |||

Trpv1
Genomic Map