siRNA Details
| Gene Symbol: Tas2r138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001451.1 | NP_001001451.1 | 1 | 514 | ||
| GenBank mRNA | AY916509.1 | - | 1 | 514 | ||
| BC128016.1 | - | 1 | 518 | 40136088 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r138| Gene Symbol: Tas2r138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001451.1 | NP_001001451.1 | 1 | 514 | ||
| GenBank mRNA | AY916509.1 | - | 1 | 514 | ||
| BC128016.1 | - | 1 | 518 | 40136088 | ||