siRNA Details
| Gene Symbol: Tpm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001491.1 | NP_001001491.1 | 8 | 1131 | ||
| GenBank mRNA | BC070421.1 | - | 8 | 1131 | 6403300 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tpm4| Gene Symbol: Tpm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001491.1 | NP_001001491.1 | 8 | 1131 | ||
| GenBank mRNA | BC070421.1 | - | 8 | 1131 | 6403300 | |