siRNA Details
Gene Symbol: 8030451A03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033210.1 | - | 2473 | |||
BB067518.2 | - | 512 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 8030451A03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033210.1 | - | 2473 | |||
BB067518.2 | - | 512 |