siRNA Details
| Gene Symbol: Dgkeos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044435.1 | - | 1211 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dgkeos| Gene Symbol: Dgkeos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044435.1 | - | 1211 | |||