siRNA Details
| Gene Symbol: Prss48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001650.1 | NP_001001650.1 | 4 | 418 | ||
| GenBank mRNA | BC116378.1 | - | 4 | 418 | 40101288 | |
| BC116379.1 | - | 4 | 418 | 40101289 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss48| Gene Symbol: Prss48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001650.1 | NP_001001650.1 | 4 | 418 | ||
| GenBank mRNA | BC116378.1 | - | 4 | 418 | 40101288 | |
| BC116379.1 | - | 4 | 418 | 40101289 | ||