siRNA Details
| Gene Symbol: ZNF425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001661.2 | NP_001001661.1 | 3 | 357 | ||
| GenBank mRNA | AK056498.1 | - | 3 | 297 | ||
| AY621067.1 | - | 3 | 297 | |||
| BC125189.1 | - | 3 | 296 | 40122655 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF425| Gene Symbol: ZNF425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001661.2 | NP_001001661.1 | 3 | 357 | ||
| GenBank mRNA | AK056498.1 | - | 3 | 297 | ||
| AY621067.1 | - | 3 | 297 | |||
| BC125189.1 | - | 3 | 296 | 40122655 | ||