siRNA Details
| Gene Symbol: ZNF782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001662.1 | NP_001001662.1 | 6 | 1829 | ||
| XM_005251742.4 | XP_005251799.1 | 5 | 1848 | |||
| XM_005251744.3 | XP_005251801.1 | 4 | 1789 | |||
| XM_011518315.2 | XP_011516617.1 | 6 | 3040 | |||
| XM_011518318.2 | XP_011516620.1 | 6 | 2027 | |||
| XM_011518320.2 | XP_011516622.1 | 5 | 1664 | |||
| XM_017014343.1 | XP_016869832.1 | 5 | 3216 | |||
| XM_017014344.1 | XP_016869833.1 | 4 | 1561 | |||
| GenBank mRNA | AK131468.1 | - | 6 | 1829 | ||
| AK302460.1 | - | 3 | 1513 | |||
| BC137073.1 | - | 6 | 1826 | 9021070 | ||

ZNF782
Genomic Map