siRNA Details
| Gene Symbol: ZC3H18-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK127438.1 | - | 2 | 2784 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZC3H18-AS1| Gene Symbol: ZC3H18-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK127438.1 | - | 2 | 2784 | ||