siRNA Details
| Gene Symbol: MIR137HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092728.1 | - | 1 | 216 | ||
| AK309618.1 | - | 1, 2 | 225 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR137HG| Gene Symbol: MIR137HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092728.1 | - | 1 | 216 | ||
| AK309618.1 | - | 1, 2 | 225 | |||