siRNA Details
| Gene Symbol: LOC400867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120405.1 | 4 | 2645 | |||
| GenBank mRNA | AK127082.1 | - | 4 | 2644 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC400867| Gene Symbol: LOC400867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120405.1 | 4 | 2645 | |||
| GenBank mRNA | AK127082.1 | - | 4 | 2644 | ||