siRNA Details
Gene Symbol: LINC00898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033377.1 | 4 | 3207 | |||
GenBank mRNA | AK128136.1 | - | 4 | 3204 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033377.1 | 4 | 3207 | |||
GenBank mRNA | AK128136.1 | - | 4 | 3204 |