siRNA Details
| Gene Symbol: LINC00487 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038369.1 | 2, 3 | 337 | |||
| GenBank mRNA | AK124409.1 | - | 2, 3 | 337 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00487| Gene Symbol: LINC00487 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038369.1 | 2, 3 | 337 | |||
| GenBank mRNA | AK124409.1 | - | 2, 3 | 337 | ||