siRNA Details
| Gene Symbol: LOC400997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122074.1 | 4 | 2537 | |||
| GenBank mRNA | AK125994.1 | - | 3 | 2355 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC400997| Gene Symbol: LOC400997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122074.1 | 4 | 2537 | |||
| GenBank mRNA | AK125994.1 | - | 3 | 2355 | ||