siRNA Details
| Gene Symbol: LINC00951 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038887.1 | 2, 3 | 2707 | |||
| GenBank mRNA | AK123643.1 | - | 2, 3 | 2707 | ||
| BC132803.1 | - | 2, 3 | 447 | |||
| BC132805.1 | - | 2, 3 | 447 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00951| Gene Symbol: LINC00951 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038887.1 | 2, 3 | 2707 | |||
| GenBank mRNA | AK123643.1 | - | 2, 3 | 2707 | ||
| BC132803.1 | - | 2, 3 | 447 | |||
| BC132805.1 | - | 2, 3 | 447 | |||