siRNA Details
| Gene Symbol: Tmem151a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001885.1 | NP_001001885.1 | 2 | 3492 | ||
| GenBank mRNA | AK139456.1 | - | 2 | 3545 | ||
| BC072634.1 | - | 2 | 3492 | 6850125 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem151a| Gene Symbol: Tmem151a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001885.1 | NP_001001885.1 | 2 | 3492 | ||
| GenBank mRNA | AK139456.1 | - | 2 | 3545 | ||
| BC072634.1 | - | 2 | 3492 | 6850125 | ||