siRNA Details
| Gene Symbol: Ephb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290753.1 | NP_001277682.1 | 16 | 3830 | ||
| NM_010142.3 | NP_034272.1 | 16 | 3827 | |||
| GenBank mRNA | BC043088.1 | - | 16 | 3751 | 6418088 | |
| BC062924.1 | - | 16 | 3775 | |||

Ephb2
Genomic Map