siRNA Details
| Gene Symbol: Olfr263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010984.1 | NP_035114.1 | 1 | 14 | ||
| GenBank mRNA | BC116918.1 | - | 1 | 93 | 40126237 | |
| BC120698.1 | - | 1 | 93 | 40129621 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr263| Gene Symbol: Olfr263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010984.1 | NP_035114.1 | 1 | 14 | ||
| GenBank mRNA | BC116918.1 | - | 1 | 93 | 40126237 | |
| BC120698.1 | - | 1 | 93 | 40129621 | ||