siRNA Details
Gene Symbol: Nrcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013150.1 | NP_037282.1 | 1638 | |||
GenBank mRNA | AY574272.1 | - | 1638 | |||
AY574273.1 | - | 1638 | ||||
AY574274.1 | - | 1638 | ||||
AY574275.1 | - | 1638 | ||||
AY574276.1 | - | 1638 | ||||
AY574277.1 | - | 1638 | ||||
U81037.1 | - | 10 | 1172 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
