siRNA Details
| Gene Symbol: OPLAH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017570.4 | NP_060040.1 | 2 | 199 | ||
| XM_011516960.1 | XP_011515262.1 | 2 | 475 | |||
| GenBank mRNA | AB122018.1 | - | 1 | 95 | ||
| BC142672.1 | - | 2 | 199 | 40148181 | ||
| BC150206.1 | - | 2 | 177 | 40148883 | ||
| CN279004.1 | - | 1 | 130 | |||

OPLAH
Genomic Map