siRNA Details
| Gene Symbol: GPR158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020752.2 | NP_065803.2 | 11 | 3245 | ||
| XM_017016452.1 | XP_016871941.1 | 8 | 2065 | |||
| GenBank mRNA | AB032962.2 | - | 3 | 1374 | ||
| AY528411.1 | - | 11 | 3245 | |||

GPR158
Genomic Map