siRNA Details
| Gene Symbol: TRAPPC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014939.3 | NP_055754.2 | 1, 2 | 528 | ||
| XM_005258233.2 | XP_005258290.1 | 1, 2 | 618 | |||
| XM_005258234.2 | XP_005258291.1 | 1, 2 | 618 | |||
| XM_006722420.3 | XP_006722483.1 | 1, 2 | 618 | |||
| XM_011525884.2 | XP_011524186.1 | 1, 2 | 618 | |||
| XM_017025616.1 | XP_016881105.1 | 1, 2 | 618 | |||
| XM_017025617.1 | XP_016881106.1 | 1, 2 | 618 | |||
| GenBank mRNA | AB023229.1 | - | 1, 2 | 221 | ||
| AK292537.1 | - | 1, 2 | 507 | |||
| AL136749.1 | - | 2, 3 | 315 | |||
| BC059380.1 | - | 1, 2 | 507 | |||
| BX642999.1 | - | 1, 2 | 74 | |||
| DA446379.1 | - | 1, 2 | 528 | |||

TRAPPC8
Genomic Map