siRNA Details
| Gene Symbol: Ypel5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027166.5 | NP_081442.1 | 3 | 1701 | ||
| GenBank mRNA | AK046281.1 | - | 3 | 1701 | ||
| AK049096.1 | - | 3 | 1702 | |||
| AK049644.1 | - | 3 | 1700 | |||
| AK137446.1 | - | 3 | 1673 | |||
| AK154461.1 | - | 3 | 1739 | |||
| AK167998.1 | - | 3 | 1719 | |||
| AK169241.1 | - | 3 | 1704 | |||
| BC085109.1 | - | 3 | 1721 | 30737658 | ||

Ypel5
Genomic Map