siRNA Details
| Gene Symbol: Tlcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291156.1 | NP_001278085.1 | 2 | 363 | ||
| NM_027249.3 | NP_081525.1 | 2 | 363 | |||
| GenBank mRNA | AK008522.1 | - | 2 | 339 | ||
| BC021945.1 | - | 2 | 334 | 5098319 | ||
| CV562896.1 | - | 2 | 364 | |||

Tlcd2
Genomic Map