siRNA Details
| Gene Symbol: Fndc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027402.3 | NP_081678.1 | 6 | 2640 | ||
| GenBank mRNA | AJ401376.1 | - | 6 | 2641 | ||
| AK137327.1 | - | 1 | 1326 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fndc5| Gene Symbol: Fndc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027402.3 | NP_081678.1 | 6 | 2640 | ||
| GenBank mRNA | AJ401376.1 | - | 6 | 2641 | ||
| AK137327.1 | - | 1 | 1326 | |||