siRNA Details
| Gene Symbol: Glipr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027450.3 | NP_081726.1 | 5 | 719 | ||
| GenBank mRNA | AK017557.1 | - | 5 | 696 | ||
| AK141459.1 | - | 5 | 710 | |||
| AK154433.1 | - | 5 | 697 | |||
| AK167595.1 | - | 5 | 719 | |||
| BC031750.1 | - | 5 | 719 | 4037002 | ||

Glipr2
Genomic Map