siRNA Details
| Gene Symbol: MORC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303256.2 | NP_001290185.1 | 6 | 745 | ||
| NM_001303257.2 | NP_001290186.1 | 6 | 745 | |||
| NM_014941.3 | NP_055756.1 | 7 | 1611 | |||
| XM_011530004.1 | XP_011528306.1 | 6 | 970 | |||
| XM_017028667.1 | XP_016884156.1 | 6 | 970 | |||
| XM_017028668.1 | XP_016884157.1 | 7 | 1065 | |||
| GenBank mRNA | AB020659.1 | - | 7 | 1525 | ||
| BC019257.1 | - | 5 | 272 | 3503651 | ||
| BC136782.1 | - | 7 | 470 | 9020772 | ||
| CR456469.1 | - | 5 | 271 | |||

MORC2
Genomic Map