siRNA Details
| Gene Symbol: Cenpu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027973.3 | NP_082249.1 | 4 | 266 | ||
| GenBank mRNA | AK006479.1 | - | 4 | 275 | ||
| AK050386.1 | - | 4 | 267 | |||
| AK081703.1 | - | 4 | 254 | |||
| AY366362.1 | - | 2 | 87 | |||
| BC094675.1 | - | 5 | 386 | |||
| BC117784.2 | - | 4 | 267 | 40092112 | ||
| BC117785.2 | - | 4 | 267 | 40092113 | ||

Cenpu
Genomic Map