siRNA Details
| Gene Symbol: Cnep1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029074.3 | NP_083350.2 | 6 | 434 | ||
| GenBank mRNA | AK017198.1 | - | 5 | 937 | ||
| AK146578.1 | - | 6 | 425 | |||
| AK217813.1 | - | 3 | 164 | |||
| BC018498.1 | - | 5 | 332 | |||
| BC118981.1 | - | 6 | 390 | |||
| BC148291.2 | - | 6 | 395 | 40103934 | ||

Cnep1r1
Genomic Map