siRNA Details
| Gene Symbol: Cnep1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029074.3 | NP_083350.2 | 6 | 496 | ||
| GenBank mRNA | AK017198.1 | - | 5 | 999 | ||
| AK146578.1 | - | 6 | 487 | |||
| BC018498.1 | - | 5 | 394 | |||
| BC118981.1 | - | 6 | 452 | |||
| BC148291.2 | - | 6 | 457 | 40103934 | ||

Cnep1r1
Genomic Map