siRNA Details
Gene Symbol: Wdr95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029440.3 | NP_083716.2 | 11, 12 | 1035 | ||
GenBank mRNA | AK087389.1 | - | 11, 12 | 1035 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wdr95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029440.3 | NP_083716.2 | 11, 12 | 1035 | ||
GenBank mRNA | AK087389.1 | - | 11, 12 | 1035 |