siRNA Details
| Gene Symbol: Olfr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146542.2 | NP_666753.2 | 1 | 28 | ||
| GenBank mRNA | BC111579.1 | - | 1 | 26 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr11| Gene Symbol: Olfr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146542.2 | NP_666753.2 | 1 | 28 | ||
| GenBank mRNA | BC111579.1 | - | 1 | 26 | ||