siRNA Details
| Gene Symbol: Foxn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183186.2 | NP_899009.2 | 6 | 1710 | ||
| GenBank mRNA | AK017346.1 | - | 5 | 1512 | ||
| AK046748.1 | - | 6 | 1985 | |||
| AK077921.1 | - | 1 | 575 | |||
| BC099971.1 | - | 6 | 1795 | 30008715 | ||

Foxn3
Genomic Map