siRNA Details
| Gene Symbol: Foxn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183186.2 | NP_899009.2 | 5, 6 | 899 | ||
| GenBank mRNA | AK017346.1 | - | 4, 5 | 702 | ||
| AK046748.1 | - | 5, 6 | 1174 | |||
| BC099971.1 | - | 5, 6 | 977 | 30008715 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxn3| Gene Symbol: Foxn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183186.2 | NP_899009.2 | 5, 6 | 899 | ||
| GenBank mRNA | AK017346.1 | - | 4, 5 | 702 | ||
| AK046748.1 | - | 5, 6 | 1174 | |||
| BC099971.1 | - | 5, 6 | 977 | 30008715 | ||