siRNA Details
| Gene Symbol: Clec4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199311.2 | NP_955015.1 | 6 | 1302 | ||
| GenBank mRNA | BC049354.1 | - | 6 | 1292 | 6310008 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec4a1| Gene Symbol: Clec4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199311.2 | NP_955015.1 | 6 | 1302 | ||
| GenBank mRNA | BC049354.1 | - | 6 | 1292 | 6310008 | |