siRNA Details
Gene Symbol: Fut7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199491.1 | NP_955785.1 | 3 | 1810 | ||
GenBank mRNA | AJ276068.2 | - | 3 | 1910 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fut7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199491.1 | NP_955785.1 | 3 | 1810 | ||
GenBank mRNA | AJ276068.2 | - | 3 | 1910 |