siRNA Details
| Gene Symbol: Lppr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201271.1 | NP_958428.1 | 4, 5 | 704 | ||
| GenBank mRNA | AY299399.1 | - | 4, 5 | 704 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lppr1| Gene Symbol: Lppr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201271.1 | NP_958428.1 | 4, 5 | 704 | ||
| GenBank mRNA | AY299399.1 | - | 4, 5 | 704 | ||