siRNA Details
| Gene Symbol: Oser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201560.2 | NP_963854.1 | 4 | 1335 | ||
| XM_006235528.2 | XP_006235590.1 | 4 | 1958 | |||
| GenBank mRNA | AJ621831.1 | - | 4 | 1339 | ||
| BC082096.1 | - | 3 | 1239 | 7134395 | ||

Oser1
Genomic Map